3-Amino-4-bromobenzoic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

A133302-100G A133302-1G A133302-25G A133302-5G
ALA133302-100GEA 1228.26 USD
ALA133302-100G ALA133302-1G ALA133302-25G ALA133302-5G
3-Amino-4-bromobenzoic acid ≥98% (by HPLC)
3-Amino-4-bromobenzoic acid
Formula: C₇H₆BrNO₂
Melting Pt: 226-230°C
Storage Temperature: Ambient
MDL Number: MFCD00579106
CAS Number: 2840-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=524f25520edbc686b9092eeb1367d0288dfe4166&vid=524f25520edbc686b9092eeb1367d0288dfe4166&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR