7-Aminodesacetoxycephalosporanic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: (6R,7R)-7-Amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Aminodeacetoxycephalosporanic acid, 7-Amino-3-methyl-3-cephem-4-carboxylic acid, 7-Aminodesacetoxycephalosporanic Acid, 7-ADCA

A133280-100G A133280-1G A133280-25G A133280-5G
ALA133280-100GEA 269.71 USD
ALA133280-100G ALA133280-1G ALA133280-25G ALA133280-5G
7-Aminodesacetoxycephalosporanic acid ≥98% (by HPLC)
7-Aminodesacetoxycephalosporanic acid

7-Aminodesacetoxycephalosporanic acid is used in the synthesis of cephalosporins and for bioconversion studies and cephalosporins.

Formula: C₈H₁₀N₂O₃S
MW: 214.24 g/mol
Melting Pt: 242°C
Storage Temperature: Refrigerator
MDL Number: MFCD00151456
CAS Number: 22252-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64367533705ba5a54f96ab035619c51241a3dcff&vid=64367533705ba5a54f96ab035619c51241a3dcff&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR