3-Aminocyclohexanecarboxylic acid (cis and trans mixture) ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

A133090-5G A133090-1G
ALA133090-5GEA 91.11 USD
ALA133090-5G ALA133090-1G
3-Aminocyclohexanecarboxylic acid (cis and trans mixture) ≥95%
3-Aminocyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₇H₁₃NO₂
MDL Number: MFCD00059562
CAS Number: 25912-50-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eba19c1d0058a5ad2085bc3ff6563529050b49b8&vid=eba19c1d0058a5ad2085bc3ff6563529050b49b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR