1-Allylnaphthalene ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC

A133081-25G A133081-5G A133081-100G A133081-1G
ALA133081-25GEA 268.76 USD
ALA133081-25G ALA133081-5G ALA133081-100G ALA133081-1G
1-Allylnaphthalene ≥97% (by GC)
1-Allylnaphthalene
Formula: C₁₃H₁₂
Boiling Pt: 133 °C
Density: 1.02 g/cm³
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00021581
CAS Number: 2489-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c651ca3c3e16e22f6abc8b595cbd4ae19906f9dd&vid=c651ca3c3e16e22f6abc8b595cbd4ae19906f9dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR