4-Amino-2-methylbenzoic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-o-toluic acid, 4-Amino-2-methylbenzoic Acid

A133080-100G A133080-1G A133080-25G A133080-5G
ALA133080-100GEA 241.21 USD
ALA133080-100G ALA133080-1G ALA133080-25G ALA133080-5G
4-Amino-2-methylbenzoic acid ≥98% (by HPLC)
4-Amino-2-methylbenzoic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 160-165°C
Storage Temperature: Ambient
MDL Number: MFCD06656133
CAS Number: 2486-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc9fc7b8e35dfcafb51e2effcf3f9d030baf4d7e&vid=bc9fc7b8e35dfcafb51e2effcf3f9d030baf4d7e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR