3-Amino-4-fluorobenzoic acid ≥97% (by titration)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A133076-1G
A133076-5G
ALA133076-1GEA
19.86
USD
ALA133076-1G
ALA133076-5G
3-Amino-4-fluorobenzoic acid ≥97% (by titration)
3-Amino-4-fluorobenzoic acid
Formula:
C₇H₆FNO₂ Melting Pt: 184 °C |
MDL Number:
MFCD00130055 CAS Number: 2365-85-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...