5-(2-Ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole ≥98% (by HPLC), Moligand™
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
Atipamezole
A132977-100MG
A132977-1G
A132977-250MG
A132977-25MG
A132977-5MG
ALA132977100MGEA
326.71
USD
ALA132977100MG
ALA132977-1G
ALA132977250MG
ALA132977-25MG
ALA132977-5MG
5-(2-Ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole ≥98% (by HPLC), Moligand™
5-(2-Ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole
Atipamezole(MPV1248) is an alpha-adrenoceptor antagonist with high affinity and selectivity for the alpha 2-receptor.
Formula:
C₁₄H₁₆N₂ MW: 212.29 g/mol Melting Pt: 129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00864502 CAS Number: 104054-27-5 |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...