Ambroxol ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-3,5-dibromo-N-(trans-4-hydroxycyclohexyl)benzylamine, Ambroxol base

A132935-1G A132935-5G
ALA132935-1GEA 112.01 USD
ALA132935-1G ALA132935-5G
Ambroxol ≥99%
Ambroxol
Formula: C₁₃H₁₈Br₂N₂O
MW: 378.1 g/mol
MDL Number: MFCD00078932
CAS Number: 18683-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4495b243eb338856a43c7d34b7d48fca06c8d0b1&vid=4495b243eb338856a43c7d34b7d48fca06c8d0b1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR