4-Amino-5-fluoro-2-methoxypyrimidine ≥98% (by GC)

Supplier: Aladdin Scientific
Warning

A132706-100G A132706-10G A132706-25G A132706-5G
ALA132706-100GEA 227.91 USD
ALA132706-100G ALA132706-10G ALA132706-25G ALA132706-5G
4-Amino-5-fluoro-2-methoxypyrimidine ≥98% (by GC)
4-Amino-5-fluoro-2-methoxypyrimidine
Formula: C₅H₆FN₃O
Melting Pt: 193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264289
CAS Number: 1993-63-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6adc832337eafce44fd987b2abe27e6314181f74&vid=6adc832337eafce44fd987b2abe27e6314181f74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR