2-Pentylpyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amylpyridine

A132572-100G A132572-1G A132572-25G A132572-5G
ALA132572-100GEA 692.46 USD
ALA132572-100G ALA132572-1G ALA132572-25G ALA132572-5G
2-Pentylpyridine ≥98% (by GC)
2-Pentylpyridine
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 207 °C
Flash Pt: 79°C
Storage Temperature: Ambient
MDL Number: MFCD00051828
CAS Number: 2294-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b08f1cd7201e695d0ffc6f2dbea4ff2ca1e852f&vid=9b08f1cd7201e695d0ffc6f2dbea4ff2ca1e852f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR