3,3,3-Trifluoro-DL-alanine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-3,3,3-trifluoropropanoic acid, H-DL-3,3,3-TriF-Ala-OH

A132515-1G A132515-250MG A132515-50MG
ALA132515-1GEA 321.96 USD
ALA132515-1G ALA132515250MG ALA132515-50MG
3,3,3-Trifluoro-DL-alanine ≥98%
3,3,3-Trifluoro-DL-alanine
Formula: C₃H₄F₃NO₂
Melting Pt: 231-234 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004263
CAS Number: 17463-43-3
EINECS: 241-478-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1962f46963dbb6556a1c0b980b1cbf74f48258e7&vid=1962f46963dbb6556a1c0b980b1cbf74f48258e7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR