N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Amino-4,6-dichloro-5-formamidopyrimidine, N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide

A132175-100G A132175-25G A132175-5G
ALA132175-100GEA 399.86 USD
ALA132175-100G ALA132175-25G ALA132175-5G
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥98% (by HPLC)
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide
Formula: C₅H₄Cl₂N₄O
MW: 207.0200 g/mol
MDL Number: MFCD04112936
CAS Number: 171887-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd12d58ff95fc0635124a93c3a806058320313fa&vid=cd12d58ff95fc0635124a93c3a806058320313fa&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR