5-Amino-4-bromo-1-methyl-1H-pyrazole ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Bromo-1-methyl-1H-pyrazol-5-amine

A132098-1G A132098-250MG
ALA132098-1GEA 44.56 USD
ALA132098-1G ALA132098250MG
5-Amino-4-bromo-1-methyl-1H-pyrazole ≥97%
5-Amino-4-bromo-1-methyl-1H-pyrazole

3-Amino-4-bromo-2-methylpyrazole (cas# 105675-85-2) is a compound useful in organic synthesis.

Formula: C₄H₆BrN₃
MW: 176.02 g/mol
Melting Pt: 96-101°C
Storage Temperature: Freezer
MDL Number: MFCD00068157
CAS Number: 105675-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13614e4f851845989f205975bd2d942311a35575&vid=13614e4f851845989f205975bd2d942311a35575&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR