4-Pentylphenol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 4-Amylphenol

A131897-100G A131897-25G A131897-5G
ALA131897-100GEA 399.86 USD
ALA131897-100G ALA131897-25G ALA131897-5G
4-Pentylphenol ≥98% (by GC)
4-Pentylphenol
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 250 °C
Melting Pt: 22 °C
Density: 0.960 g/cm³
Flash Pt: 133°C
Storage Temperature: Ambient
MDL Number: MFCD00020211
CAS Number: 14938-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bdbb118630a196b60270cc088af340dae605f0a&vid=3bdbb118630a196b60270cc088af340dae605f0a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR