ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: ABD-F, 4-Fluoro-7-sulfobenzofurazan, ammonium salt, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulphonamide, 4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide

A131562-100MG A131562-25MG
ALA131562100MGEA 276.36 USD
ALA131562100MG ALA131562-25MG
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98% (by HPLC)
ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole]

Highly reactive fluorogenic reagent for the labeling of thiols for HPLC. ABD-F is nonfluorescent until reacted with thiols and therefore can be used to quantitate thiols in solution, as well as thiols separated by HPLC or TLC.

Formula: C₆H₄FN₃O₃S
MW: 217.17 g/mol
Melting Pt: 146 °C
Storage Temperature: Freezer
MDL Number: MFCD00036687
CAS Number: 91366-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db256daa902b6a87c7b1120f5c507083236a703d&vid=db256daa902b6a87c7b1120f5c507083236a703d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR