Anthracene-9,10-dipropionic acid disodium salt ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Sodium 3,3'-(anthracene-9,10-diyl)dipropanoate, ADPA

A131550-25MG A131550-50MG
ALA131550-25MGEA 360.91 USD
ALA131550-25MG ALA131550-50MG
Anthracene-9,10-dipropionic acid disodium salt ≥98% (by HPLC)
Anthracene-9,10-dipropionic acid disodium salt

A naphthalene as singlet oxygen sensitizer and a self assembly building block.

Formula: C₂₀H₁₆Na₂O₄
MW: 366.32 g/mol
MDL Number: MFCD00467617
CAS Number: 82767-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e25f0af94a01b066bb1adba2cdb89179eb55e965&vid=e25f0af94a01b066bb1adba2cdb89179eb55e965&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR