9,10-Anthracenediyl-bis(methylene)dimalonic acid ≥90%
Supplier: Aladdin Scientific
|
Warning
|
Product Describtion:Reagent for the assay of singlett oxygen; it has better characteristics than 9,10-anthracenediyl-bis-dipropionic acid; In addition to the emission maximum at 407 nm, there are lower maxima at 431, 457 and 485 nmA useful O2 determining agent.Product Application:9,10-Anthracenediyl-bis(methylene)dimalonic acid is widely used as a singlet oxygen detector probe. 9,10-Anthracenediyl-bis(methylene)dimalonic acid is suitable for assessing the photodynamic effect of curcumin Vibrio parahaemolyticus, which is a significant cause of bacterial diarrhea
Formula:
C₂₂H₁₈O₈ Storage Temperature: Ambient |
MDL Number:
MFCD03095464 CAS Number: 307554-62-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...