Albendazole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: [5-(Propylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester, Methyl [5-(Propylthio)benzimidazol-2-yl]carbamate

A131023-100G A131023-25G A131023-50G A131023-5G
ALA131023-100GEA 264.01 USD
ALA131023-100G ALA131023-25G ALA131023-50G ALA131023-5G
Albendazole ≥98%
Albendazole

Binds to tubulin and inhibits microtubule assembly.

Formula: C₁₂H₁₅N₃O₂S
MW: 265.33 g/mol
Melting Pt: 208-210°C
MDL Number: MFCD00083232
CAS Number: 54965-21-8
EINECS: 259-414-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49cf1f2c50a0b95e428a03d496fd846521e485a0&vid=49cf1f2c50a0b95e428a03d496fd846521e485a0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR