2-Amino-5-bromobenzonitrile ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
4-Bromo-2-cyanoaniline
A130043-100G
A130043-1G
A130043-250MG
A130043-25G
A130043-5G
ALA130043-100GEA
1013.56
USD
ALA130043-100G
ALA130043-1G
ALA130043250MG
ALA130043-25G
ALA130043-5G
2-Amino-5-bromobenzonitrile ≥98% (by GC)
2-Amino-5-bromobenzonitrile
Melting point:96-100℃2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.
Formula:
C₇H₅BrN₂ Melting Pt: 94.0-98.0°C Storage Temperature: Freezer |
MDL Number:
MFCD00158946 CAS Number: 39263-32-6 EINECS: 254-387-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...