2-Amino-5-mercapto-1,3,4-thiadiazole ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Amino-1,3,4-thiadiazole-2-thiol

A130041-100G A130041-25G
ALA130041-100GEA 55.96 USD
ALA130041-100G ALA130041-25G
2-Amino-5-mercapto-1,3,4-thiadiazole ≥98% (by HPLC)
2-Amino-5-mercapto-1,3,4-thiadiazole
Formula: C₂H₃N₃S₂
MW: 133.2 g/mol
Melting Pt: 235°C
Storage Temperature: Ambient
MDL Number: MFCD00003108
CAS Number: 2349-67-9
EINECS: 219-078-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d5c18ad1b403201d46cf62e19125bf2256295d5&vid=5d5c18ad1b403201d46cf62e19125bf2256295d5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR