2-(Methylsulfonyl)ethanamine hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-(Methylsulphonyl)ethanamine hydrochloride, 2-Aminoethyl Methyl sulphone Hydrochloride, 2-Aminoethyl methyll sulphone hydrochloride, 2-(Methylsulphonyl)ethanamine Hydrochloride

A130035-1G A130035-25G A130035-5G
ALA130035-1GEA 10.36 USD
ALA130035-1G ALA130035-25G ALA130035-5G
2-(Methylsulfonyl)ethanamine hydrochloride ≥98%
2-(Methylsulphonyl)ethanamine hydrochloride
Formula: C₃H₁₀ClNO₂S
MW: 159.6400 g/mol
Melting Pt: 168.0-172.0°C
MDL Number: MFCD03840162
CAS Number: 104458-24-4
EINECS: 450-260-8, 690-962-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fd84aef478baad1f9ef3601137343ca04216683&vid=2fd84aef478baad1f9ef3601137343ca04216683&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR