5-Aminosalicylic acid ≥99%, Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Mesalamine is an anti-inflammatory compound.5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read
Formula:
C₇H₇NO₃ Melting Pt: 265°C Flash Pt: 280 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007877 CAS Number: 89-57-6 EINECS: 201-919-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...