Amorolfine hydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A129973-100MG A129973-10G A129973-1G A129973-250MG A129973-5G
ALA129973100MGEA 98.71 USD
ALA129973100MG ALA129973-10G ALA129973-1G ALA129973250MG ALA129973-5G
Amorolfine hydrochloride ≥98% (by HPLC)
Amorolfine hydrochloride

A broad-based antifungal agent which blocks ergosterol biosynthesis.

Formula: C₂₁H₃₅NO
Melting Pt: 204-208°C
Storage Temperature: Refrigerator
MDL Number: MFCD00903738
CAS Number: 78613-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a633d45dfe8a3b0a81b93e16e688e96a2024ccf4&vid=a633d45dfe8a3b0a81b93e16e688e96a2024ccf4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR