TC-S 7010 ≥98% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Benzamide, TC‐S7010, TCS7010, Aurora A Inhibitor I, N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide, N-(2-Chlorophenyl)-4-(2-(4-(2-(4-ethylpiperazin-1-yl)-2-oxoethyl)phenylamino)-5-fluoropyrimidin-4-ylamino)benzamide, N-(2-Chlorophenyl)-4-((2-((4-(2-(4-ethylpiperazin-1-yl)-2-oxoethyl)phenyl)amino)-5-fluoropyrimidin-4-yl)amino)benzamide

A129933-100MG A129933-10MG A129933-25MG A129933-50MG A129933-5MG
ALA129933100MGEA 892.91 USD
ALA129933100MG ALA129933-10MG ALA129933-25MG ALA129933-50MG ALA129933-5MG
TC-S 7010 ≥98% (by HPLC), Moligand™
TC-S 7010

Aurora A Inhibitor I is a novel, potent, and selective inhibitor of Aurora A with IC50 of 3.4 nM in a cell-free assay. It is 1000-fold more selective for Aurora A than Aurora B.

Formula: C₃₁H₃₁ClFN₇O₂
Storage Temperature: Freezer
MDL Number: MFCD16495816
CAS Number: 1158838-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44f23ce0f1bd37caf4387eb49b15be6cbcc3d9dc&vid=44f23ce0f1bd37caf4387eb49b15be6cbcc3d9dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR