AMG-900 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A129924-10MG A129924-1MG A129924-25MG A129924-50MG A129924-5MG
ALA129924-10MGEA 233.61 USD
ALA129924-10MG ALA129924-1MG ALA129924-25MG ALA129924-50MG ALA129924-5MG
AMG-900 ≥98%, Moligand™
AMG-900
Formula: C₂₈H₂₁N₇OS
Storage Temperature: Freezer
MDL Number: MFCD18633194
CAS Number: 945595-80-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65204fb75bdd92d72f5a4d00aec3246f8f553a8b&vid=65204fb75bdd92d72f5a4d00aec3246f8f553a8b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR