Ricobendazole ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Albendazole sulphoxide, N-[6-(propylsulphinyl)-1H-benzimidazol-2-yl]-carbamic acid, methyl ester, Albendazole Oxide, Methyl (6-(propylsulphinyl)-1H-benzo[d]imidazol-2-yl)carbamate
A129901-1G
A129901-25G
A129901-5G
ALA129901-1GEA
9.41
USD
ALA129901-1G
ALA129901-25G
ALA129901-5G
Ricobendazole ≥98%
Ricobendazole
Albendazole Oxide is a tubulin polymerization or assembly inhibitorRicobendazole is a methylcarbamate benzimidazole with a broad-spectrum anthelmintic activity. Ricobendazole is a key metabolite of albendazole;It has been shown to induce apoptosis in human cancer cell line HT-29, possibly by arresting the cell cycle at the G2/M phase.
Formula:
C₁₂H₁₅N₃O₃S Melting Pt: 226 °C |
MDL Number:
MFCD00797922 CAS Number: 54029-12-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...