Adenine hydrochloride hemihydrate ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Purin-6-amine hydrochloride hemihydrate, 1H-Purin-6-amine hydrochloride, 7H-Purin-6-amine hydrochloride hemihydrate, 6-Aminopurine Hydrochloride, Adenine Hydrochloride Hemihydrate, 6-Aminopurine hydrochloride hemihydrate, 7H-Purin-6-amine hydrochloride hydrate (2:2:1)

A129851-100G A129851-25G
ALA129851-100GEA 984.11 USD
ALA129851-100G ALA129851-25G
Adenine hydrochloride hemihydrate ≥99%
Adenine hydrochloride hemihydrate

Adenine HCl is a hydrochloride salt form of adenine which is a purine derivative and a nucleobase with a variety of roles in biochemistryAdenine is a purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro. It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).

Formula: C₅H₅N₅·HCl·½H₂O
MW: 180.60 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11046306
CAS Number: 6055-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1693cf1697b08baa085dd70649635cc9d9a474fd&vid=1693cf1697b08baa085dd70649635cc9d9a474fd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR