AM-251 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-(2,4-Dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-1-piperidinyl-1H-pyrazole-3-carboxamide, 1-(2,4-Dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide, AM 251, AM251, AM-251

A129844-25MG A129844-5MG
ALA129844-25MGEA 231.71 USD
ALA129844-25MG ALA129844-5MG
AM-251 ≥98%, Moligand™
AM-251

AM251 block the inhibitory effects of endocannabinoids and synthetic cannabinoid agonists on transmitter release through an action at presynaptic cannabinoid 1 receptors in brain.An inverse agonist of the CB1 receptor; AM251, a CB1 cannabinoid receptor antagonist, has been used in a study to determine its interaction with hippocampal neurons to enhance spatial memory in mice.

Formula: C₂₂H₂₁Cl₂IN₄O
MW: 555.2 g/mol
Melting Pt: 196-198°C
Storage Temperature: Refrigerator
MDL Number: MFCD01861181
CAS Number: 183232-66-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc21e827b81bcd8c641c3f0d131ac1ce3f1d0eaf&vid=fc21e827b81bcd8c641c3f0d131ac1ce3f1d0eaf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR