AM 1241 ≥98%, Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
AM1241, (R,S)-3-(2-Iodo-5-nitrobenzoyl)-1-(1-methyl-2-piperidinylmethyl)-1H-indole, (R,S)-AM1241
A129838-5MG
A129838-10MG
A129838-1MG
A129838-25MG
ALA129838-5MGEA
97.76
USD
ALA129838-5MG
ALA129838-10MG
ALA129838-1MG
ALA129838-25MG
AM 1241 ≥98%, Moligand™
AM 1241
AM-1241 is a selective cannabinoid CB2 receptor agonist with Ki of 3.4 nM, exhibits 82-fold selectivity over CB1 receptor.(R,S)-AM1241, a selective CB2 cannabinoid receptor agonist, has been used to study neuropathic pain in animal models
Formula:
C₂₂H₂₂N₃O₃I Storage Temperature: Freezer |
CAS Number:
444912-48-5 EINECS: 636-937-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...