Acebutolol hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-(3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl)butanamide, Acebutolol HCl, Acebutolol hydrochloride

A129820-100MG A129820-50MG
ALA129820100MGEA 43.61 USD
ALA129820100MG ALA129820-50MG
Acebutolol hydrochloride ≥98%
Acebutolol hydrochloride
Formula: C₁₈H₂₈N₂O₄·HCl
MW: 372.89 g/mol
Melting Pt: 143-145°C
Storage Temperature: Freezer
MDL Number: MFCD00078860
CAS Number: 34381-68-5
EINECS: 251-980-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426921b6fe0eee942621e7f71b453d1a8151f0d3&vid=426921b6fe0eee942621e7f71b453d1a8151f0d3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR