Acarbose ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Prandase, Acarbose Hydrate, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay

A129816-1G A129816-250MG
ALA129816-1GEA 85.41 USD
ALA129816-1G ALA129816250MG
Acarbose ≥98%, Moligand™
Acarbose

Acarbose, also known as BAY g 5421, is an α-glucosidase inhibitor that prevents absorption of sucrose and maltose. This compound has been found to delay digestion of complex disaccharides and carbohydrates. Further studies suggest that this compound can stimulate phosphorylase kinase (PhK) by binding to the glucoamylase-like domain new the amino termini and causing an alteration in the structure.An α-glucosidase inhibitorApplication:Acarbose, a derivitized maltotriose antidiabetic drug, is a reversible α-glucosidase inhibitor used in medical studies to control the effects of Type II diabetes. Acarbose is often used as a reference to which other α-glucosidase inhibitors are compared.

Formula: C₂₅H₄₃NO₁₈
MW: 645.61 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00869592
CAS Number: 56180-94-0
EINECS: 260-030-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=938bcc28a822a0567abf1b2f9168d5486fd0b81f&vid=938bcc28a822a0567abf1b2f9168d5486fd0b81f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR