Alfuzosin hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)(methyl)amino]propyl]tetrahydro-2-furancarboxamide Hydrochloride, 4-Amino-6,7-dimethoxy-2-[methyl[3-[(tetrahydro-2-furoyl)amino]propyl]amino]quinazoline Hydrochloride, Alfuzosin HCl, N-(3-((4-Amino-6,7-dimethoxyquinazolin-2-yl)(methyl)amino)propyl)tetrahydrofuran-2-carboxamide hydrochloride

A129804-100MG A129804-25MG A129804-50MG A129804-5G A129804-1G A129804-250MG
ALA129804100MGEA 366.61 USD
ALA129804100MG ALA129804-25MG ALA129804-50MG ALA129804-5G ALA129804-1G ALA129804250MG
Alfuzosin hydrochloride ≥98%
Alfuzosin hydrochloride

Alfuzosin HCl is an alpha1 receptor antagonist used to treat benign prostatic hyperplasia (BPH).An α 1 adrenoceptor antagonist.

Formula: C₁₉H₂₇N₅O₄
MW: 425.91 g/mol
Melting Pt: 229 °C
MDL Number: MFCD00879135
CAS Number: 81403-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cb8a62fa1a734b9a921aa7f6044619c382c6703&vid=0cb8a62fa1a734b9a921aa7f6044619c382c6703&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR