(1R,2R)-2-(4'-(3-Phenylureido)-[1,1'-biphenyl]-4-carbonyl)cyclopentanecarboxylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: (1R,2R)-2-[4-[4-(phenylcarbamoylamino)phenyl]benzoyl]cyclopentane-1-carboxylic acid, A 922500, (1R,2R)-2-[[4'-[[Phenylamino)carbonyl]amino] [1,1'-biphenyl]-4-yl]carbonyl]cyclopentanecarboxylic acid, A922500, DGAT-1 Inhibitor 4a

A129800-10MG A129800-1MG A129800-50MG A129800-5MG
ALA129800-10MGEA 165.21 USD
ALA129800-10MG ALA129800-1MG ALA129800-50MG ALA129800-5MG
(1R,2R)-2-(4'-(3-Phenylureido)-[1,1'-biphenyl]-4-carbonyl)cyclopentanecarboxylic acid ≥98%
(1R,2R)-2-(4'-(3-Phenylureido)-[1,1'-biphenyl]-4-carbonyl)cyclopentanecarboxylic acid

Product DescriptionDGAT-1 inhibitor A 922500 (A-922500) reduces serum triglyceride levels from baseline at all doses tested; however, this is only statistically significant at the 3 mg/kg dose, which lowers serum triglycerides by 53%. Similarly, the 3 mg/kg dose of A 922500 significantly reduces serum FFA concentrations by 55% and total cholesterol by 25%. DGAT-1 inhibition has no significant effect on body weight at any dose tested. Although A 922500 dpes not significantly affect LDL-cholesterol or HDL-cholesterol individually, the serum LDL/HDL-cholesterol ratio is significantly improved by A 922500 at 0.3 and 3 mg/kg. Similar to the dyslipidemic hamster, treatment with 3 mg/kg A 922500 significantly reduces serum triglyceride concentrations (39%). FFA levels significantly increase over the 14-day period in vehicle-treated animals. This increase is inhibited in a dose-dependent manner by A 922500 such that FFA concentrations are 32% lower after 14 days of treatment with the DGAT-1 inhibitor at 3 mg/kg, compared with the vehicle group (p < 0.05). HDL-cholesterol is significantly increased from baseline levels by A 922500 at 0.3 and 3 mg/kg; however, this is only significantly increased compared with vehicle at the 3 mg/kg dose. Body weight significantly increases over the 2-week period in vehicle-treated rats, and this is not affected by A 922500. LDL-cholesterol is significantly reduced in the vehicle treated group. DGAT-1 inhibition does not further reduce LDL-cholesterol and has no effect on total cholesterol.Product ApplicationA922500 has been used in the inhibition of diacylglycerol acyltransferase (DGAT) in epithelial human breast cancer cell line (MDA-MB-231), digitonin-permeabilized human cancer cells, human myotubes and macrophages.

Formula: C₂₆H₂₄N₂O₄
MW: 428.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12828780
CAS Number: 959122-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96f9c859c12c53f581bc7ad1a7ee2862413816dc&vid=96f9c859c12c53f581bc7ad1a7ee2862413816dc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR