Abacavir sulfate ≥99%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A129792-10MG
A129792-1G
A129792-250MG
A129792-50MG
A129792-5G
ALA129792-10MGEA
46.46
USD
ALA129792-10MG
ALA129792-1G
ALA129792250MG
ALA129792-50MG
ALA129792-5G
Abacavir sulfate ≥99%
Abacavir sulfate
Abacavir is a commonly used nucleoside analogue with potent antiviral activity against HIV-1.A nucleoside reverse transcriptase inhibitor.
Formula:
C₂₈H₃₈N₁₂O₆S Storage Temperature: Refrigerator |
MDL Number:
MFCD04112763 CAS Number: 188062-50-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...