5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: AICAR, 5-Amino-1-(β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine

A129749-1G A129749-250MG A129749-50MG
ALA129749-1GEA 754.21 USD
ALA129749-1G ALA129749250MG ALA129749-50MG
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98%, Moligand™
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside

Acadesine results in accumulation of ZMP, which mimics the stimulating effect of AMP on AMPK and AMPK kinase.An inhibitor of the transcription of PPARα, the coactivation of PPARα.

Formula: C₉H₁₄N₄O₅
MW: 258.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00869751
CAS Number: 2627-69-2
EINECS: 220-097-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3e6aff3567e15b3cb97a93f5e93e42cb62b9dd6&vid=a3e6aff3567e15b3cb97a93f5e93e42cb62b9dd6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR