Azathioprine ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 6-((1-Methyl-4-nitro-1H-imidazol-5-yl)thio)-9H-purine, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

A129740-1G A129740-25G A129740-5G
ALA129740-1GEA 34.11 USD
ALA129740-1G ALA129740-25G ALA129740-5G
Azathioprine ≥98%, Moligand™
Azathioprine

Azathioprine is a drug that suppresses the immune system and is used in organ transplantation and autoimmune diseaseAn immunosuppressive antimetabolite that interferes with the synthesis of purines.

Formula: C₉H₇N₇O₂S
MW: 277.26 g/mol
Melting Pt: 244°C
Storage Temperature: Freezer
MDL Number: MFCD00069203
CAS Number: 446-86-6
EINECS: 207-175-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a5d08e887e0ca9b3f31039a23a12de9d9c82046&vid=3a5d08e887e0ca9b3f31039a23a12de9d9c82046&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR