Abiraterone Acetate ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

A129731-5G A129731-5MG A129731-100MG A129731-1G A129731-250MG A129731-25G A129731-25MG
ALA129731-5GEA 3289.76 USD
ALA129731-5G ALA129731-5MG ALA129731100MG ALA129731-1G ALA129731250MG ALA129731-25G ALA129731-25MG
Abiraterone Acetate ≥99%, Moligand™
Abiraterone Acetate

Abiraterone Acetate is an acetate salt form of Abiraterone which is a steroidal cytochrome CYP17 inhibitor with IC50 of 72 nM.A steroidal inhibitor of CYP17A1 in androgen biosynthesis

Formula: C₂₆H₃₃NO₂
Melting Pt: 147 °C
Storage Temperature: Freezer
MDL Number: MFCD00934213
CAS Number: 154229-18-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7008d7afcaf08eb5be8041e52c0bd8e14bfa39f&vid=e7008d7afcaf08eb5be8041e52c0bd8e14bfa39f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR