Amitriptyline hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Elavil hydrochloride, 5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, Amitriptyline Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-5-ylidene)-N,N-dimethyl-1-propanamine hydrochloride, Endep hydrochloride, Levate hydrochloride

A129730-100G A129730-1G A129730-25G A129730-5G
ALA129730-100GEA 1600.66 USD
ALA129730-100G ALA129730-1G ALA129730-25G ALA129730-5G
Amitriptyline hydrochloride ≥98%
Amitriptyline hydrochloride

Amitriptyline inhibits serotonin receptor, norepinephrine receptor, 5-HT4, 5-HT2 and sigma 1 receptor with IC50 of 3.45 nM, 13.3 nM, 7.31 nM, 235 nM and 287 nM, respectively.Amitriptyline hydrochloride is a tricyclic antidepressant drug. Amitriptyline hydrochloride inhibits the norepinephrine and serotonin transporters but has a high affinity for α1-adrenergic, serotonin and muscarinic acetylcholine receptors.

Formula: C₂₀H₂₃N
MW: 313.87 g/mol
Melting Pt: 195 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012537
CAS Number: 549-18-8
EINECS: 208-964-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ee6d0179d721196a2b009d799ddf1a643f0b1e9&vid=2ee6d0179d721196a2b009d799ddf1a643f0b1e9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR