N-(4-((6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yl)oxy)benzo[d]thiazol-2-yl)acetamide ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: N-[4-[[6-[4-(Trifluoromethyl)phenyl]-4-pyrimidinyl]oxy]-2-benzothiazolyl]acetamide, AMG-517, AMG 517, AMG 517, N-(4-(6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yloxy)benzo[d]thiazol-2-yl)acetamide

A129715-50MG A129715-10MG A129715-1MG A129715-25MG
ALA129715-50MGEA 422.66 USD
ALA129715-50MG ALA129715-10MG ALA129715-1MG ALA129715-25MG
N-(4-((6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yl)oxy)benzo[d]thiazol-2-yl)acetamide ≥99%, Moligand™
N-(4-((6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-yl)oxy)benzo[d]thiazol-2-yl)acetamide
Formula: C₂₀H₁₃F₃N₄O₂S
MW: 430.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD14584859
CAS Number: 659730-32-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c553e17f55e9e31f2c7dc8ab50e9bb71ed157acb&vid=c553e17f55e9e31f2c7dc8ab50e9bb71ed157acb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR