Almotriptan Malate ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
3-[2-(Dimethylamino)ethyl]-5-(pyrrolidin-1-ylsulphonylmethyl)-1H-indole Malate
A129703-10MG
A129703-1G
A129703-250MG
A129703-50MG
ALA129703-10MGEA
95.86
USD
ALA129703-10MG
ALA129703-1G
ALA129703250MG
ALA129703-50MG
Almotriptan Malate ≥98%
Almotriptan Malate
Almotriptan Malate is a selective 5-hydroxytryptamine1B/1D (5-HT1B/1D) receptor agonist, used for the treatment of Migraine attacks in adultsA serotonin 5HT1B /1D-receptor agonist used for treatment of migraines.
Formula:
C₁₇H₂₅N₃O₂S·C₄H₆O₅ MW: 469.55 g/mol Melting Pt: 162 °C Storage Temperature: Freezer |
MDL Number:
MFCD08067740 CAS Number: 181183-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...