6,7-Dichloro-5,10-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Anagrelide HCl, Agrylin hydrochloride, 6,7-Dichloro-1,5-dihydroimidazo[2,1-b]quinazolin-2(3H)-one monohydrochloride, BL4162A, Anagrelide hydrochloride, Xagrid hydrochloride

A129685-1G A129685-250MG A129685-50MG
ALA129685-1GEA 3340.11 USD
ALA129685-1G ALA129685250MG ALA129685-50MG
6,7-Dichloro-5,10-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride ≥98%
6,7-Dichloro-5,10-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride

A potent type III phosphodiesterase inhibitor

Formula: C₁₀H₈Cl₃N₃O
MW: 292.55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01720337
CAS Number: 58579-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c2e6ebae22b058754420eb5e3c8898a9e9f89f&vid=82c2e6ebae22b058754420eb5e3c8898a9e9f89f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR