Avanafil ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-[(3-Chloro-4-methoxybenzyl)amino]-2-[(2S)-2-(hydroxymethyl)pyrrolidin-1-yl]-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide, Avanfil, (S)-4-((3-Chloro-4-methoxybenzyl)amino)-2-(2-(hydroxymethyl)pyrrolidin-1-yl)-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide

A129676-1G A129676-250MG A129676-25G A129676-50MG A129676-5G
ALA129676-1GEA 1200.71 USD
ALA129676-1G ALA129676250MG ALA129676-25G ALA129676-50MG ALA129676-5G
Avanafil ≥99%, Moligand™
Avanafil

Avanafil is a highly selective PDE5 inhibitor with IC50 of 5.2 nM, >121-fold selectivity over other PDEs.

Formula: C₂₃H₂₆ClN₇O₃
MW: 483.951 g/mol
Melting Pt: 165°C
Storage Temperature: Freezer
MDL Number: MFCD11977961
CAS Number: 330784-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5ae8d4dbed2ad9e2a19755c98a1f541e67b8a2a&vid=c5ae8d4dbed2ad9e2a19755c98a1f541e67b8a2a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR