Atracurium besylate (mixture of isomers) 98

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Atracurium Besylate, 51W89, BW-33A, 2,2'-[1,5-Pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium] Dibenzenesulfonate (mixture of isomers), 2,2'-[1,5-Pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium] Dibenzenesulphonate, Tracrium

A129655-1G A129655-250MG
ALA129655-1GEA 32.21 USD
ALA129655-1G ALA129655250MG
Atracurium besylate (mixture of isomers) 98
Atracurium besylate (mixture of isomers)

Atracurium Besylate is a neuromuscular blocking agent with ED95 of 0.2 mg/kg.Atracurium Besylate , a specialty product for proteomics research

Formula: C₆₅H₈₂N₂O₁₈S₂
MW: 1243.49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00797403
CAS Number: 64228-81-5
EINECS: 264-743-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e81631b56adbaf622b340d6514a99163bd97106d&vid=e81631b56adbaf622b340d6514a99163bd97106d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR