AZ 628 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: AZ 628, 3-(2-cyanopropan-2-yl)-N-[4-methyl-3-[(3-methyl-4-oxoquinazolin-6-yl)amino]phenyl]benzamide, AZ-628, AZ628, 3-(2-Cyanopropan-2-yl)-N-(4-methyl-3-((3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl)amino)phenyl)benzamide

A129605-10MG A129605-1MG A129605-25MG A129605-50MG A129605-5MG
ALA129605-10MGEA 112.96 USD
ALA129605-10MG ALA129605-1MG ALA129605-25MG ALA129605-50MG ALA129605-5MG
AZ 628 ≥98%, Moligand™
AZ 628
Formula: C₂₇H₂₅N₅O₂
MW: 451.53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17392577
CAS Number: 878739-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2fc0579b421836fcf930c676f8f8a0642ca2172&vid=b2fc0579b421836fcf930c676f8f8a0642ca2172&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR