Amoxapine ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A129596-1G A129596-250MG A129596-50MG A129596-5G
ALA129596-1GEA 115.81 USD
ALA129596-1G ALA129596250MG ALA129596-50MG ALA129596-5G
Amoxapine ≥98%, Moligand™
Amoxapine

Amoxapine is a tricyclic antidepressant known to inhibit reuptake of neuronal transmitter. The drug helps in controlling agitation and anxiety along with depression.Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Application:Amoxapine may be used as a pharmaceutical reference standard for the determination of the analyte in drug dosage forms by ultraviolet-spectrophotometric method.These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Formula: C₁₇H₁₆ClN₃O
Melting Pt: 178.0-182.0 °C
Storage Temperature: Freezer
MDL Number: MFCD00069210
CAS Number: 14028-44-5
EINECS: 237-867-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b69afd7791473e3fc68a1b5016f241c54ce98c51&vid=b69afd7791473e3fc68a1b5016f241c54ce98c51&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR