Amiodarone hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Amiodarone HCl, (2-Butylbenzofuran-3-yl)(4-(2-(diethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride)

A129574-25G A129574-5G A129574-100G A129574-1G
ALA129574-25GEA 500.56 USD
ALA129574-25G ALA129574-5G ALA129574-100G ALA129574-1G
Amiodarone hydrochloride ≥98%
Amiodarone hydrochloride

An inhibitor of 1,4-dihydropyridine binding to L-type Ca2+ channels

Formula: C₂₅H₂₉I₂NO₃
Melting Pt: 156-163°C
Storage Temperature: Refrigerator
MDL Number: MFCD00069204
CAS Number: 19774-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c1b3906a24f812e67a3df0c5eb9a403596d0896&vid=5c1b3906a24f812e67a3df0c5eb9a403596d0896&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR