Ambroxol hydrochloride ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride, Ambroxol HCl, trans-4-((2-Amino-3,5-dibromobenzyl)amino)cyclohexanol hydrochloride

A129556-1G A129556-25G A129556-5G
ALA129556-1GEA 18.91 USD
ALA129556-1G ALA129556-25G ALA129556-5G
Ambroxol hydrochloride ≥99%
Ambroxol hydrochloride

AmbroxolHCl is a potent inhibitor of the neuronal Na+ channels, inhibits TTX-resistant Na+ currents with IC50 of 35.2 μM and 22.5 μM for tonic and phasic block, inhibits TTX-sensitive Na+ currents with IC50 of 100 μM. Phase 3.An expectorant antioxidant and antiinflammatory agent

Formula: C₁₃H₁₈Br₂N₂O.HCl
MW: 414.57 g/mol
Melting Pt: 235°C
MDL Number: MFCD00078932
CAS Number: 23828-92-4
EINECS: 245-899-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c25814fd1272479aecf61bc16e823f2f667b0303&vid=c25814fd1272479aecf61bc16e823f2f667b0303&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR