Amiloride hydrochloride dihydrate ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A129545-1G A129545-200MG A129545-5G
ALA129545-1GEA 195.61 USD
ALA129545-1G ALA129545200MG ALA129545-5G
Amiloride hydrochloride dihydrate ≥98%
Amiloride hydrochloride dihydrate

Amiloride HCl dihydrate is a potent epithelial sodium channel blocker.An epithelial sodium channel blocker

Formula: C₆H₈ClN₇O·HCl·2H₂O
Melting Pt: 293.5°C
Storage Temperature: Freezer
MDL Number: MFCD00058197
CAS Number: 17440-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dc51f2be2c9bd18974de74287f0cf47decfe07e&vid=9dc51f2be2c9bd18974de74287f0cf47decfe07e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR