AS-604850 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A129517-25MG A129517-100MG
ALA129517-25MGEA 140.51 USD
ALA129517-25MG ALA129517100MG
AS-604850 ≥98%, Moligand™
AS-604850

AS-604850 is a selective, ATP-competitive PI3Kγ inhibitor with IC50 of 250 nM, over 80-fold selectivity for PI3Kγ than PI3Kδ/β, and 18-fold more selective for PI3Kγ than PI3Kα.An ATP-competitive inhibitor of PI 3-kinase γ.

Formula: C₁₁H₅F₂NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD19705528
CAS Number: 648449-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0617029bcb934e442d9be238f77cef41a889607&vid=d0617029bcb934e442d9be238f77cef41a889607&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR