Sodium-2-(2-amino-3-benzoylphenyl)acetate hydrate ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Amfenac Sodium Hydrate, Amfenac sodium, Sodium 2-(2-amino-3-benzoylphenyl)acetate hydrate, Fenazox, Amfenac Sodium Monohydrate

A129507-1G A129507-250MG
ALA129507-1GEA 1062.01 USD
ALA129507-1G ALA129507250MG
Sodium-2-(2-amino-3-benzoylphenyl)acetate hydrate ≥99%
Sodium-2-(2-amino-3-benzoylphenyl)acetate hydrate

An antibacterial agent.

Formula: C₁₅H₁₄NNaO₄
MW: 295.2657 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00941383
CAS Number: 61618-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71ab6a3954156b218e50cdc663309b7de5fc5596&vid=71ab6a3954156b218e50cdc663309b7de5fc5596&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR